Assay Details
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011933.1 | NP_001011933.1 | 8 - 9 | 1019 | 94 | |
XM_006230020.2 | XP_006230082.1 | 9 - 10 | 1103 | 94 | ||
GenBank mRNA | BC082015.1 | - | 8 - 9 | 1019 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
