Assay Details
Gene Symbol: Mtch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106488.2 | NP_001099958.2 | 754 | 208 | ||
GenBank mRNA | BC158687.1 | - | 705 | 8375010 | 208 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
