Assay Details
Gene Symbol: Srf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109302.1 | NP_001102772.1 | 2 - 3 | 1093 | 76 | |
XM_006244557.2 | XP_006244619.1 | 2 - 3 | 1096 | 76 | ||
XM_006244558.2 | XP_006244620.1 | 2 - 3 | 1097 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
