Assay Details
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232168.1 | XP_006232230.1 | 1 - 2 | 286 | 70 | |
XM_008760873.1 | XP_008759095.1 | 1 - 2 | 286 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232168.1, XM_008760873.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
