Assay Details
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232168.1 | XP_006232230.1 | 4 - 5 | 540 | 146 | |
XM_008760873.1 | XP_008759095.1 | 4 - 5 | 540 | 146 | ||
XM_008760874.1 | XP_008759096.1 | 3 - 4 | 3171 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
