Assay Details
Gene Symbol: D2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106926.1 | NP_001100396.1 | 3 - 4 | 737 | 67 | |
XM_006245520.2 | XP_006245582.1 | 3 - 4 | 653 | 67 | ||
XM_006245521.2 | XP_006245583.1 | 3 - 4 | 511 | 67 | ||
XM_006245522.2 | XP_006245584.1 | 3 - 4 | 506 | 67 | ||
XM_008767365.1 | XP_008765587.1 | 3 - 4 | 768 | 67 | ||
GenBank mRNA | BC166559.1 | - | 2 - 3 | 410 | 7312233 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
