Assay Details
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108546.1 | NP_001102016.1 | 1 - 2 | 263 | 144 | |
XM_006232168.1 | XP_006232230.1 | 1 - 2 | 286 | 290 | ||
XM_008760873.1 | XP_008759095.1 | 1 - 2 | 286 | 290 | ||
XM_008760875.1 | XP_008759097.1 | 1 - 2 | 254 | 144 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108546.1, XM_006232168.1, XM_008760873.1, XM_008760875.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
