Assay Details
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108546.1 | NP_001102016.1 | 2 - 3 | 295 | 107 | |
XM_008760875.1 | XP_008759097.1 | 2 - 3 | 286 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
