Assay Details
Gene Symbol: Pcdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761025.1 | XP_008759247.1 | 2 - 3 | 3147 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
