Assay Details
Gene Symbol: Sox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105952.1 | NP_001099422.1 | 12 - 13 | 1694 | 62 | |
XM_006249762.2 | XP_006249824.1 | 11 - 12 | 1358 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P073954 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P073959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173461 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073959 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073954 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073954 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073959 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P073959 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P073954 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P302068 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56594 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10767691 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767680 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767677 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767689 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767686 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767687 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767683 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767688 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767679 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767684 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767685 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767681 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767682 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10767675 | - | - |
Rat Genome 230 2.0 Array | 1374682_at | - | - |
Rat Genome 230 2.0 Array | 1374682_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374682_at | - | - |
Affymetrix HT Rat Focus Array | 1374682_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300001844.rn.1 | TC1300000288.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001847.rn.1 | TC1300000288.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001848.rn.1 | TC1300000288.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300001845.rn.1 | TC1300000288.rn.1 | RTA TC |
Rat Expression Set 230 | 1374682_at | - | - |
Rat Expression Set 230 | 1374682_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767675 | - | - |
Rat Genome U34 Set | rc_AI013074_at | - | - |
Rat Genome U34 Set | rc_AI013074_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21556158 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374539 | - | - |
