Assay Details
Gene Symbol: Stk25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_184049.1 | NP_908938.1 | 7 - 8 | 932 | 90 | |
GenBank mRNA | AY346152.1 | - | 7 - 8 | 932 | 90 | |
BC087092.1 | - | 7 - 8 | 878 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
