Assay Details
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108346.2 | NP_001101816.1 | 5 - 6 | 477 | 72 | |
XM_006249926.2 | XP_006249988.1 | 6 - 7 | 700 | 72 | ||
XM_006249927.2 | XP_006249989.1 | 6 - 7 | 4318 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
