Assay Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021762.1 | NP_068530.1 | 4 - 5 | 478 | 74 | |
GenBank mRNA | AF262356.1 | - | 4 - 5 | 478 | 74 | |
AY325258.1 | - | 553 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
