Assay Details
Gene Symbol: Cdh20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012748.1 | NP_001012766.1 | 9 - 10 | 1640 | 114 | |
GenBank mRNA | AB121033.1 | - | 9 - 10 | 1640 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
