Assay Details
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031068.1 | NP_112330.1 | 9 - 10 | 1032 | 86 | |
GenBank mRNA | AB292042.1 | - | 9 - 10 | 956 | 86 | |
AB292043.1 | - | 9 - 10 | 945 | 86 | ||
BC098014.1 | - | 9 - 10 | 1032 | 7454624 | 86 | |
FQ212256.1 | - | 9 - 10 | 1234 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
