Assay Details
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013223.1 | NP_001013241.1 | 8 - 9 | 812 | 105 | |
XM_006242417.2 | XP_006242479.1 | 8 - 9 | 822 | 105 | ||
XM_006242418.2 | XP_006242480.1 | 8 - 9 | 810 | 105 | ||
XM_006242419.2 | XP_006242481.1 | 8 - 9 | 732 | 105 | ||
XM_006242420.2 | XP_006242482.1 | 8 - 9 | 810 | 105 | ||
XM_006242421.2 | XP_006242483.1 | 8 - 9 | 822 | 105 | ||
XM_006242422.2 | XP_006242484.1 | 8 - 9 | 810 | 105 | ||
XM_008765770.1 | XP_008763992.1 | 8 - 9 | 744 | 105 | ||
GenBank mRNA | BC078906.1 | - | 8 - 9 | 812 | 105 | |
BC078909.1 | - | 7 - 8 | 639 | 105 | ||
FQ231388.1 | - | 4 - 5 | 353 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
