Assay Details
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109445.1 | NP_001102915.1 | 5 - 6 | 459 | 78 | |
XM_006240399.2 | XP_006240461.1 | 5 - 6 | 433 | 78 | ||
GenBank mRNA | BC158833.1 | - | 5 - 6 | 401 | 78 | |
FJ179410.1 | - | 4 - 5 | 336 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
