Assay Details
Gene Symbol: Dhrs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014119.2 | NP_001014141.2 | 1 - 2 | 255 | 78 | |
XM_006247045.2 | XP_006247107.1 | 1 - 2 | 376 | 78 | ||
XM_008768069.1 | XP_008766291.1 | 1 - 2 | 361 | 78 | ||
GenBank mRNA | BC088444.1 | - | 1 - 2 | 249 | 78 | |
DY311021.1 | - | 1 - 2 | 255 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088444.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
