Assay Details
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053428.1 | NP_445880.1 | 2 - 3 | 264 | 70 | |
XM_006257592.1 | XP_006257654.1 | 2 - 3 | 532 | 70 | ||
XM_008773572.1 | XP_008771794.1 | 3 - 4 | 440 | 70 | ||
XM_008773573.1 | XP_008771795.1 | 3 - 4 | 502 | 70 | ||
GenBank mRNA | AF271786.1 | - | 2 - 3 | 264 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
