Assay Details
Gene Symbol: Akap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053665.1 | NP_446117.1 | 6 - 7 | 2282 | 88 | |
XM_006247067.2 | XP_006247129.1 | 7 - 8 | 2455 | 88 | ||
XM_006247068.1 | XP_006247130.1 | 7 - 8 | 2451 | 88 | ||
XM_008767930.1 | XP_008766152.1 | 7 - 8 | 2510 | 88 | ||
GenBank mRNA | AF068202.1 | - | 6 - 7 | 2282 | 88 | |
BC081753.1 | - | 8 - 9 | 2534 | 88 | ||
BC097298.1 | - | 9 - 10 | 2626 | 88 | ||
BC129090.1 | - | 9 - 10 | 2605 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
