Assay Details
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242423.2 | XP_006242485.1 | 8 - 9 | 824 | 65 | |
XM_006242424.2 | XP_006242486.1 | 8 - 9 | 1244 | 65 | ||
XM_006242425.2 | XP_006242487.1 | 8 - 9 | 812 | 65 | ||
XM_006242426.2 | XP_006242488.1 | 8 - 9 | 824 | 65 | ||
XM_006242427.2 | XP_006242489.1 | 8 - 9 | 812 | 65 | ||
XM_008765771.1 | XP_008763993.1 | 8 - 9 | 734 | 65 | ||
GenBank mRNA | BC097410.1 | - | 9 - 10 | 929 | 65 | |
FQ216778.1 | - | 8 - 9 | 703 | 65 | ||
FQ220695.1 | - | 4 - 5 | 392 | 65 | ||
FQ220700.1 | - | 2 - 3 | 167 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
