Assay Details
Gene Symbol: Zfand6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007630.1 | NP_001007631.1 | 5 - 6 | 510 | 65 | |
XM_006229499.2 | XP_006229561.1 | 7 - 8 | 815 | 65 | ||
XM_006229500.2 | XP_006229562.1 | 5 - 6 | 495 | 65 | ||
XM_006229502.2 | XP_006229564.1 | 6 - 7 | 638 | 65 | ||
XM_006229504.1 | XP_006229566.1 | 6 - 7 | 649 | 65 | ||
XM_006229505.2 | XP_006229567.1 | 5 - 6 | 503 | 65 | ||
XM_006229506.2 | XP_006229568.1 | 5 - 6 | 500 | 65 | ||
XM_008759611.1 | XP_008757833.1 | 5 - 6 | 2040 | 65 | ||
GenBank mRNA | BC076394.1 | - | 5 - 6 | 510 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
