Assay Details
Gene Symbol: Pcf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759657.1 | XP_008757879.1 | 3 - 4 | 824 | 94 | |
XM_341883.6 | XP_341884.4 | 3 - 4 | 824 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
