Assay Details
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025689.1 | NP_001020860.1 | 8 - 9 | 852 | 141 | |
GenBank mRNA | BC097427.1 | - | 8 - 9 | 852 | 141 | |
BC158645.1 | - | 8 - 9 | 800 | 8363877 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
