Assay Details
Gene Symbol: Crat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004085.2 | NP_001004085.1 | 2 - 3 | 516 | 67 | |
XM_006233823.1 | XP_006233885.1 | 3 - 4 | 1036 | 67 | ||
XM_008761663.1 | XP_008759885.1 | 3 - 4 | 519 | 67 | ||
XM_008761664.1 | XP_008759886.1 | 2 - 3 | 385 | 67 | ||
GenBank mRNA | AJ620886.1 | - | 2 - 3 | 294 | 67 | |
BC083616.1 | - | 2 - 3 | 516 | 7133847 | 67 | |
FQ230824.1 | - | 3 - 4 | 631 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
