Assay Details
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106513.1 | NP_001099983.1 | 2 - 3 | 634 | 159 | |
XM_008762162.1 | XP_008760384.1 | 2 - 3 | 579 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
