Assay Details
Gene Symbol: Pdia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017319.1 | NP_059015.1 | 11 - 12 | 1416 | 64 | |
GenBank mRNA | BC062393.1 | - | 11 - 12 | 1453 | 64 | |
D63378.1 | - | 11 - 12 | 1416 | 64 | ||
FQ211627.1 | - | 1515 | 64 | |||
X12355.1 | - | 11 - 12 | 1409 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
