Assay Details
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235015.2 | XP_006235077.1 | 1 - 2 | 232 | 68 | |
XM_006235016.2 | XP_006235078.1 | 2 - 3 | 1604 | 68 | ||
XM_006235017.2 | XP_006235079.1 | 1 - 2 | 139 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
