Assay Details
Gene Symbol: Disp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107759.2 | NP_001101229.2 | 1 - 2 | 219 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107759.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
