Assay Details
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760610.1 | XP_008758832.1 | 12 - 13 | 1824 | 68 | |
XM_008760611.1 | XP_008758833.1 | 11 - 12 | 1785 | 68 | ||
XM_008760612.1 | XP_008758834.1 | 13 - 14 | 4504 | 68 | ||
XM_008760613.1 | XP_008758835.1 | 12 - 13 | 4368 | 68 | ||
XM_008760614.1 | XP_008758836.1 | 12 - 13 | 1820 | 68 | ||
XM_008760615.1 | XP_008758837.1 | 11 - 12 | 1781 | 68 | ||
XM_008760616.1 | XP_008758838.1 | 8 - 9 | 3974 | 68 | ||
XM_008760617.1 | XP_008758839.1 | 8 - 9 | 855 | 68 | ||
GenBank mRNA | FQ232558.1 | - | 11 - 12 | 1404 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
