Assay Details
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760610.1 | XP_008758832.1 | 13 - 14 | 1932 | 140 | |
XM_008760611.1 | XP_008758833.1 | 12 - 13 | 1893 | 140 | ||
XM_008760612.1 | XP_008758834.1 | 14 - 15 | 4612 | 140 | ||
XM_008760613.1 | XP_008758835.1 | 13 - 14 | 4476 | 140 | ||
XM_008760614.1 | XP_008758836.1 | 13 - 14 | 1928 | 140 | ||
XM_008760615.1 | XP_008758837.1 | 12 - 13 | 1889 | 140 | ||
XM_008760616.1 | XP_008758838.1 | 9 - 10 | 4082 | 140 | ||
XM_008760617.1 | XP_008758839.1 | 9 - 10 | 963 | 140 | ||
GenBank mRNA | FQ232558.1 | - | 12 - 13 | 1512 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
