Assay Details
Gene Symbol: Sept11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107208.2 | NP_001100678.1 | 778 | 72 | ||
GenBank mRNA | BC167769.1 | - | 909 | 7456582 | 72 | |
EU711414.1 | - | 778 | 72 | |||
EU711415.1 | - | 778 | 72 | |||
EU711416.1 | - | 778 | 72 | |||
EU711417.1 | - | 778 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
