Assay Details
Gene Symbol: Hhatl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106868.1 | NP_001100338.1 | 3 - 4 | 336 | 61 | |
XM_006244090.2 | XP_006244152.1 | 3 - 4 | 1238 | 61 | ||
XM_006244091.2 | XP_006244153.1 | 3 - 4 | 417 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
