Assay Details
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106131.1 | NP_001099601.1 | 5 - 6 | 909 | 153 | |
XM_006254942.2 | XP_006255004.1 | 5 - 6 | 2014 | 153 | ||
XM_006254943.2 | XP_006255005.1 | 5 - 6 | 2015 | 153 | ||
XM_006254944.2 | XP_006255006.1 | 5 - 6 | 2016 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
