Assay Details
Gene Symbol: U2af2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748763.3 | XP_003748811.1 | 9 - 10 | 1041 | 69 | |
XM_006228320.2 | XP_006228382.1 | 9 - 10 | 1041 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
