Assay Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106924.1 | NP_001100394.1 | 4 - 5 | 858 | 119 | |
XM_006245373.2 | XP_006245435.1 | 5 - 6 | 1007 | 119 | ||
XM_006245374.2 | XP_006245436.1 | 5 - 6 | 1150 | 119 | ||
XM_006245375.2 | XP_006245437.1 | 5 - 6 | 909 | 119 | ||
GenBank mRNA | FQ225617.1 | - | 1 - 2 | 174 | 119 | |
FQ232683.1 | - | 1 - 2 | 174 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
