Assay Details
Gene Symbol: Pde6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108806.1 | NP_001102276.1 | 3 - 4 | 400 | 128 | |
XM_006245413.2 | XP_006245475.1 | 3 - 4 | 448 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
