Assay Details
Gene Symbol: Serpine2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019197.1 | NP_062070.1 | 5 - 6 | 1010 | 73 | |
XM_006245161.2 | XP_006245223.1 | 5 - 6 | 1673 | 73 | ||
XM_006245162.2 | XP_006245224.1 | 6 - 7 | 1137 | 73 | ||
XM_008767212.1 | XP_008765434.1 | 5 - 6 | 997 | 73 | ||
GenBank mRNA | CF110312.1 | - | 1 - 2 | 168 | 73 | |
FQ228974.1 | - | 3 - 4 | 579 | 73 | ||
M17784.1 | - | 4 - 5 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
