Assay Details
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007713.1 | NP_001007714.1 | 10 - 11 | 845 | 144 | |
XM_008767238.1 | XP_008765460.1 | 10 - 11 | 3217 | 144 | ||
XM_008767239.1 | XP_008765461.1 | 11 - 12 | 868 | 144 | ||
GenBank mRNA | BC079087.1 | - | 10 - 11 | 845 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
