Assay Details
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007713.1 | NP_001007714.1 | 1 - 2 | 254 | 65 | |
XM_008767238.1 | XP_008765460.1 | 1 - 2 | 2626 | 65 | ||
GenBank mRNA | BC079087.1 | - | 1 - 2 | 254 | 7123233 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
