Assay Details
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007713.1 | NP_001007714.1 | 3 - 4 | 420 | 65 | |
XM_008767238.1 | XP_008765460.1 | 3 - 4 | 2792 | 65 | ||
XM_008767239.1 | XP_008765461.1 | 4 - 5 | 443 | 65 | ||
GenBank mRNA | BC079087.1 | - | 3 - 4 | 420 | 7123233 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
