Assay Details
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014219.1 | NP_001014241.1 | 11 - 12 | 2264 | 69 | |
XM_006245248.2 | XP_006245310.1 | 10 - 11 | 2161 | 69 | ||
GenBank mRNA | BC081973.1 | - | 11 - 12 | 2264 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
