Assay Details
Gene Symbol: Sphkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127492.1 | NP_001120964.1 | 7 - 8 | 917 | 69 | |
XM_006245230.2 | XP_006245292.1 | 7 - 8 | 1089 | 69 | ||
XM_006245231.2 | XP_006245293.1 | 7 - 8 | 1490 | 69 | ||
XM_006245233.2 | XP_006245295.1 | 6 - 7 | 1009 | 69 | ||
XM_006245234.2 | XP_006245296.1 | 7 - 8 | 1088 | 69 | ||
XM_008767251.1 | XP_008765473.1 | 6 - 7 | 1411 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
