Assay Details
Gene Symbol: Prss56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750730.3 | XP_003750778.1 | 9 - 10 | 1252 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
