Assay Details
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108808.1 | NP_001102278.1 | 1 - 2 | 577 | 73 | |
GenBank mRNA | FQ219916.1 | - | 108 | 73 | ||
FQ220431.1 | - | 151 | 73 | |||
FQ226325.1 | - | 120 | 73 | |||
FQ231706.1 | - | 120 | 73 | |||
FQ232529.1 | - | 120 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108808.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
