Assay Details
Gene Symbol: Eif4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC158817.1 | - | 38 | 7374173 | 61 | |
FQ233479.1 | - | 59 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC158817.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
