Assay Details
Gene Symbol: Upp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030025.1 | NP_001025196.1 | 1 - 2 | 75 | 80 | |
XM_006251483.2 | XP_006251545.1 | 2 - 3 | 444 | 80 | ||
XM_006251484.2 | XP_006251546.1 | 2 - 3 | 350 | 80 | ||
XM_006251485.2 | XP_006251547.1 | 2 - 3 | 294 | 80 | ||
XM_006251487.2 | XP_006251549.1 | 2 - 3 | 444 | 80 | ||
XM_008770270.1 | XP_008768492.1 | 2 - 3 | 268 | 80 | ||
GenBank mRNA | BC099752.1 | - | 1 - 2 | 75 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC099752.1, NM_001030025.1, XM_006251483.2, XM_006251484.2, XM_006251485.2, XM_006251487.2, XM_008770270.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
