Assay Details
Gene Symbol: Upp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030025.1 | NP_001025196.1 | 3 - 4 | 273 | 58 | |
XM_006251483.2 | XP_006251545.1 | 4 - 5 | 642 | 58 | ||
XM_006251484.2 | XP_006251546.1 | 4 - 5 | 548 | 58 | ||
XM_006251485.2 | XP_006251547.1 | 4 - 5 | 492 | 58 | ||
XM_006251487.2 | XP_006251549.1 | 4 - 5 | 639 | 58 | ||
XM_008770270.1 | XP_008768492.1 | 4 - 5 | 466 | 58 | ||
GenBank mRNA | BC099752.1 | - | 3 - 4 | 273 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
