Assay Details
Gene Symbol: Upp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251483.2 | XP_006251545.1 | 1 - 2 | 373 | 76 | |
XM_006251487.2 | XP_006251549.1 | 1 - 2 | 373 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006251483.2, XM_006251487.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
