Assay Details
Gene Symbol: Igfbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012588.2 | NP_036720.2 | 4 - 5 | 991 | 82 | |
GenBank mRNA | BF281457.1 | - | 3 - 4 | 454 | 82 | |
M31837.1 | - | 4 - 5 | 944 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
